Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258411.2 | NP_001245340.1 | 10 - 11 | 1279 | 133 |
NM_001258412.2 | NP_001245341.1 | 9 - 10 | 1174 | 133 | |
NM_001258413.2 | NP_001245342.1 | 10 - 11 | 1225 | 133 | |
NM_001258414.1 | NP_001245343.1 | 9 - 10 | 1019 | 133 | |
NM_052944.4 | NP_443176.2 | 10 - 11 | 1235 | 133 | |
XM_005255080.2 | XP_005255137.1 | 11 - 12 | 1263 | 133 | |
XM_005255082.4 | XP_005255139.1 | 5 - 6 | 669 | 133 | |
XM_006721015.2 | XP_006721078.1 | 9 - 10 | 1243 | 133 | |
XM_011545725.2 | XP_011544027.2 | 10 - 11 | 1406 | 133 | |
XM_011545728.1 | XP_011544030.1 | 2 - 3 | 202 | 133 | |
XM_017022900.1 | XP_016878389.1 | 11 - 12 | 1579 | 133 | |
XM_017022901.1 | XP_016878390.1 | 11 - 12 | 2289 | 133 | |
XM_017022902.1 | XP_016878391.1 | 11 - 12 | 1418 | 133 | |
XM_017022903.1 | XP_016878392.1 | 12 - 13 | 1428 | 133 | |
XM_017022904.1 | XP_016878393.1 | 12 - 13 | 1598 | 133 | |
XM_017022905.1 | XP_016878394.1 | 10 - 11 | 1474 | 133 | |
XM_017022906.1 | XP_016878395.1 | 11 - 12 | 1439 | 133 | |
XM_017022907.1 | XP_016878396.1 | 10 - 11 | 1057 | 133 | |
XM_017022908.1 | XP_016878397.1 | 10 - 11 | 1116 | 133 | |
XM_017022909.1 | XP_016878398.1 | 9 - 10 | 952 | 133 | |
GenBank mRNA | AF292385.1 | - | 9 - 10 | 1003 | 133 |
AJ305237.1 | - | 9 - 10 | 1021 | 133 | |
AK125267.1 | - | 12 - 13 | 1351 | 133 | |
AK295427.1 | - | 9 - 10 | 1043 | 133 | |
AK298373.1 | - | 9 - 10 | 1020 | 133 | |
AK303691.1 | - | 10 - 11 | 1147 | 133 | |
AY044906.1 | - | 10 - 11 | 1235 | 133 | |
BC049385.1 | - | 9 - 10 | 889 | 133 | |
BC057780.1 | - | 10 - 11 | 1094 | 133 |