Assay Design
Gene Transcripts
- Gene Symbol
- ADORA1
- Entrez Gene ID
-
134
- Gene Name
- adenosine A1 receptor
- Gene Aliases
-
RDC7
- Chromosome Location
-
Chr.1: 203127705 - 203167405 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
L22214.1 |
- |
1 - 2 |
85 |
74 |
View Full Product Details