Assay Design



Gene Transcripts

Gene Symbol
YTHDF1
Entrez Gene ID
54915
Gene Name
YTH N6-methyladenosine RNA binding protein 1
Gene Aliases
C20orf21
Chromosome Location
Chr.20: 63195429 - 63216186 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017798.3 NP_060268.2 2 - 3 347 91
GenBank mRNA AK095795.1 - 2 - 3 65 91
BC050284.1 - 2 - 3 347 91

View Full Product Details