Assay Design



Gene Transcripts

Gene Symbol
XPO7
Entrez Gene ID
23039
Gene Name
exportin 7
Gene Aliases
EXP7, RANBP16
Chromosome Location
Chr.8: 21919669 - 22006585 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015024.4 NP_055839.3 10 - 11 1209 79
XM_017013244.1 XP_016868733.1 10 - 11 1463 79
XM_017013245.1 XP_016868734.1 10 - 11 1230 79
XM_017013246.1 XP_016868735.1 10 - 11 1432 79
XM_017013247.1 XP_016868736.1 10 - 11 1623 79
GenBank mRNA AF064729.1 - 10 - 11 1119 79
AK291472.1 - 10 - 11 1124 79
AK299848.1 - 6 - 7 645 79
BC030785.1 - 10 - 11 1208 79

View Full Product Details