Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007055.3 | NP_008986.2 | 12 - 13 | 1779 | 79 |
GenBank mRNA | AF021351.1 | - | 13 - 14 | 1646 | 79 |
AK026023.1 | - | 3 - 4 | 353 | 79 | |
AK300329.1 | - | 12 - 13 | 1759 | 79 | |
AY091460.1 | - | 5 - 6 | 554 | 79 | |
BC041089.1 | - | 12 - 13 | 1740 | 79 |