Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007055.3 | NP_008986.2 | 14 - 15 | 2032 | 80 |
GenBank mRNA | AF021351.1 | - | 15 - 16 | 1899 | 80 |
AK026023.1 | - | 5 - 6 | 606 | 80 | |
AK300329.1 | - | 14 - 15 | 2012 | 80 | |
AY091460.1 | - | 7 - 8 | 807 | 80 | |
BC041089.1 | - | 14 - 15 | 1993 | 80 |