Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007055.3 | NP_008986.2 | 23 - 24 | 3204 | 65 |
GenBank mRNA | AF021351.1 | - | 24 - 25 | 3071 | 65 |
AK026023.1 | - | 14 - 15 | 1778 | 65 | |
AK300329.1 | - | 23 - 24 | 3184 | 65 | |
BC041089.1 | - | 23 - 24 | 3165 | 65 |