Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007055.3 | NP_008986.2 | 28 - 29 | 3899 | 61 |
GenBank mRNA | AF021351.1 | - | 29 - 30 | 3766 | 61 |
AK026023.1 | - | 19 - 20 | 2473 | 61 | |
AK300329.1 | - | 29 - 30 | 3816 | 61 | |
AY091462.1 | - | 4 - 5 | 493 | 61 | |
BC014399.1 | - | 5 - 6 | 605 | 61 | |
BC041089.1 | - | 28 - 29 | 3860 | 61 |