Assay Design



Gene Transcripts

Gene Symbol
THOP1
Entrez Gene ID
7064
Gene Name
thimet oligopeptidase 1
Gene Aliases
EP24.15, MEPD_HUMAN, MP78, TOP
Chromosome Location
Chr.19: 2785460 - 2813604 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003249.4 NP_003240.1 10 - 11 1846 77
XM_011528228.2 XP_011526530.1 7 - 8 2793 77
GenBank mRNA AK093392.1 - 1 - 2 964 77
AK300552.1 - 7 - 8 1328 77
BC000135.2 - 10 - 11 1772 77
BC000583.2 - 8 - 9 1351 77
BC002391.2 - 10 - 11 1798 77
BC013878.2 - 10 - 11 1762 77
U29366.1 - 10 - 11 1730 77
Z50115.1 - 10 - 11 1770 77

View Full Product Details