Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001447.2 | NP_001438.1 | 7 - 8 | 4588 | 117 |
XM_006714761.3 | XP_006714824.1 | 8 - 9 | 5159 | 117 | |
XM_011537600.2 | XP_011535902.1 | 10 - 11 | 4927 | 117 | |
XM_011537603.2 | XP_011535905.1 | 8 - 9 | 4860 | 117 | |
XM_017009224.1 | XP_016864713.1 | 10 - 11 | 4929 | 117 | |
XM_017009225.1 | XP_016864714.1 | 9 - 10 | 4771 | 117 | |
XM_017009226.1 | XP_016864715.1 | 8 - 9 | 5164 | 117 | |
XM_017009227.1 | XP_016864716.1 | 8 - 9 | 5163 | 117 | |
GenBank mRNA | AF231022.1 | - | 7 - 8 | 4588 | 117 |