Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099737.2 | NP_001093207.1 | 8 - 9 | 918 | 78 |
NM_032332.3 | NP_115708.1 | 6 - 7 | 661 | 78 | |
NR_029375.1 | 8 - 9 | 901 | 78 | ||
GenBank mRNA | AK074525.1 | - | 8 - 9 | 891 | 78 |
AK223195.1 | - | 6 - 7 | 611 | 78 | |
AK314164.1 | - | 8 - 9 | 895 | 78 | |
BC005870.2 | - | 6 - 7 | 594 | 78 |