Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001149.3 | NP_001140.2 | 2 - 3 | 794 | 91 |
NM_001204403.1 | NP_001191332.1 | 25 - 26 | 2932 | 91 | |
NM_001204404.1 | NP_001191333.1 | 25 - 26 | 3175 | 91 | |
NM_001320874.1 | NP_001307803.1 | 24 - 25 | 3189 | 91 | |
NM_020987.4 | NP_066267.2 | 24 - 25 | 3189 | 91 | |
XM_005269715.3 | XP_005269772.1 | 25 - 26 | 3076 | 91 | |
XM_017016111.1 | XP_016871600.1 | 25 - 26 | 3076 | 91 | |
XM_017016122.1 | XP_016871611.1 | 24 - 25 | 3022 | 91 | |
XM_017016130.1 | XP_016871619.1 | 24 - 25 | 3022 | 91 | |
XM_017016132.1 | XP_016871621.1 | 24 - 25 | 3022 | 91 | |
XM_017016133.1 | XP_016871622.1 | 24 - 25 | 3121 | 91 | |
XM_017016135.1 | XP_016871624.1 | 24 - 25 | 3121 | 91 | |
XM_017016139.1 | XP_016871628.1 | 24 - 25 | 3022 | 91 | |
XM_017016140.1 | XP_016871629.1 | 24 - 25 | 3022 | 91 | |
XM_017016150.1 | XP_016871639.1 | 24 - 25 | 3022 | 91 | |
GenBank mRNA | AK092527.1 | - | 2 - 3 | 799 | 91 |
AK126851.1 | - | 6 - 7 | 877 | 91 | |
AK292927.1 | - | 2 - 3 | 680 | 91 | |
AK295661.1 | - | 25 - 26 | 2882 | 91 | |
AL136710.1 | - | 2 - 3 | 453 | 91 | |
BX537917.1 | - | 25 - 26 | 2803 | 91 | |
BX648574.1 | - | 25 - 26 | 2897 | 91 | |
U13616.1 | - | 24 - 25 | 2935 | 91 |