Assay Design



Gene Transcripts

Gene Symbol
ANK3
Entrez Gene ID
288
Gene Name
ankyrin 3, node of Ranvier (ankyrin G)
Gene Aliases
ANKYRIN-G, MRT37
Chromosome Location
Chr.10: 60026298 - 60733526 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204403.1 NP_001191332.1 4 - 5 507 64
NM_001204404.1 NP_001191333.1 3 - 4 696 64
NM_001320874.1 NP_001307803.1 3 - 4 764 64
NM_020987.4 NP_066267.2 3 - 4 764 64
XM_005269715.3 XP_005269772.1 3 - 4 597 64
XM_006717796.3 XP_006717859.1 3 - 4 597 64
XM_006717802.3 XP_006717865.1 3 - 4 597 64
XM_011539708.2 XP_011538010.1 3 - 4 597 64
XM_011539709.2 XP_011538011.1 3 - 4 597 64
XM_017016110.1 XP_016871599.1 3 - 4 597 64
XM_017016111.1 XP_016871600.1 3 - 4 597 64
XM_017016112.1 XP_016871601.1 3 - 4 597 64
XM_017016113.1 XP_016871602.1 3 - 4 597 64
XM_017016114.1 XP_016871603.1 3 - 4 696 64
XM_017016115.1 XP_016871604.1 3 - 4 426 64
XM_017016116.1 XP_016871605.1 3 - 4 597 64
XM_017016117.1 XP_016871606.1 3 - 4 597 64
XM_017016118.1 XP_016871607.1 3 - 4 597 64
XM_017016119.1 XP_016871608.1 3 - 4 597 64
XM_017016120.1 XP_016871609.1 3 - 4 597 64
XM_017016121.1 XP_016871610.1 3 - 4 597 64
XM_017016122.1 XP_016871611.1 3 - 4 597 64
XM_017016123.1 XP_016871612.1 3 - 4 597 64
XM_017016124.1 XP_016871613.1 3 - 4 597 64
XM_017016125.1 XP_016871614.1 3 - 4 597 64
XM_017016126.1 XP_016871615.1 3 - 4 597 64
XM_017016127.1 XP_016871616.1 3 - 4 597 64
XM_017016128.1 XP_016871617.1 3 - 4 597 64
XM_017016129.1 XP_016871618.1 3 - 4 597 64
XM_017016130.1 XP_016871619.1 3 - 4 597 64
XM_017016131.1 XP_016871620.1 3 - 4 597 64
XM_017016132.1 XP_016871621.1 3 - 4 597 64
XM_017016133.1 XP_016871622.1 3 - 4 696 64
XM_017016134.1 XP_016871623.1 3 - 4 597 64
XM_017016135.1 XP_016871624.1 3 - 4 696 64
XM_017016136.1 XP_016871625.1 3 - 4 597 64
XM_017016137.1 XP_016871626.1 3 - 4 597 64
XM_017016138.1 XP_016871627.1 3 - 4 597 64
XM_017016139.1 XP_016871628.1 3 - 4 597 64
XM_017016140.1 XP_016871629.1 3 - 4 597 64
XM_017016141.1 XP_016871630.1 3 - 4 597 64
XM_017016142.1 XP_016871631.1 3 - 4 597 64
XM_017016143.1 XP_016871632.1 3 - 4 597 64
XM_017016144.1 XP_016871633.1 3 - 4 597 64
XM_017016145.1 XP_016871634.1 3 - 4 597 64
XM_017016146.1 XP_016871635.1 3 - 4 597 64
XM_017016147.1 XP_016871636.1 3 - 4 597 64
XM_017016148.1 XP_016871637.1 3 - 4 479 64
XM_017016149.1 XP_016871638.1 3 - 4 696 64
XM_017016150.1 XP_016871639.1 3 - 4 597 64
GenBank mRNA AB209308.1 - 3 - 4 649 64
AK295661.1 - 3 - 4 403 64
BP245303.1 - 4 - 5 507 64
BX537917.1 - 4 - 5 378 64
BX648574.1 - 4 - 5 472 64
DB237893.1 - 3 - 4 331 64
U13616.1 - 3 - 4 510 64

View Full Product Details