Assay Design



Gene Transcripts

Gene Symbol
ZFP28
Entrez Gene ID
140612
Gene Name
ZFP28 zinc finger protein
Gene Aliases
mkr5
Chromosome Location
Chr.19: 56536791 - 56559213 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308440.1 NP_001295369.1 5 - 6 756 77
NM_020828.1 NP_065879.1 5 - 6 756 77
XM_011526462.2 XP_011524764.1 5 - 6 752 77
XM_011526463.2 XP_011524765.2 5 - 6 1485 77
GenBank mRNA AB037852.1 - 5 - 6 756 77
AF507045.1 - 5 - 6 756 77
AK289583.1 - 5 - 6 741 77
BC028416.2 - 5 - 6 751 77
BC099903.1 - 5 - 6 751 77
BC127002.1 - 5 - 6 741 77
BC127003.1 - 5 - 6 741 77

View Full Product Details