Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018439.3 | NP_060909.1 | 10 - 11 | 1003 | 77 |
XM_017025832.1 | XP_016881321.1 | 10 - 11 | 1256 | 77 | |
GenBank mRNA | AB026264.1 | - | 10 - 11 | 930 | 77 |
AF208694.1 | - | 10 - 11 | 891 | 77 | |
AK292533.1 | - | 10 - 11 | 1003 | 77 | |
AK312727.1 | - | 10 - 11 | 978 | 77 | |
BC034016.1 | - | 10 - 11 | 951 | 77 | |
BC036074.1 | - | 9 - 10 | 2126 | 77 |