Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018439.3 | NP_060909.1 | 4 - 5 | 399 | 133 |
XM_017025832.1 | XP_016881321.1 | 4 - 5 | 652 | 133 | |
GenBank mRNA | AB026264.1 | - | 4 - 5 | 326 | 133 |
AK292533.1 | - | 4 - 5 | 399 | 133 | |
AK312727.1 | - | 4 - 5 | 374 | 133 | |
BC034016.1 | - | 4 - 5 | 347 | 133 | |
BC036074.1 | - | 3 - 4 | 1522 | 133 |