Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001184993.1 | NP_001171922.1 | 4 - 5 | 474 | 97 |
NM_031277.2 | NP_112567.2 | 4 - 5 | 474 | 97 | |
XM_006719846.3 | XP_006719909.1 | 4 - 5 | 768 | 97 | |
XM_006719849.2 | XP_006719912.1 | 4 - 5 | 485 | 97 | |
XM_006719852.2 | XP_006719915.1 | 3 - 4 | 363 | 97 | |
XM_011535152.2 | XP_011533454.1 | 8 - 9 | 2393 | 97 | |
XM_011535155.2 | XP_011533457.1 | 7 - 8 | 1129 | 97 | |
XM_011535156.2 | XP_011533458.1 | 8 - 9 | 2393 | 97 | |
XM_011535157.2 | XP_011533459.1 | 8 - 9 | 2393 | 97 | |
XM_011535158.2 | XP_011533460.1 | 8 - 9 | 2393 | 97 | |
XM_011535159.2 | XP_011533461.1 | 8 - 9 | 2393 | 97 | |
XM_011535160.2 | XP_011533462.1 | 8 - 9 | 2393 | 97 | |
XM_017020676.1 | XP_016876165.1 | 4 - 5 | 437 | 97 | |
GenBank mRNA | AF285602.1 | - | 4 - 5 | 438 | 97 |
AF285603.1 | - | 4 - 5 | 438 | 97 | |
AK302405.1 | - | 3 - 4 | 363 | 97 | |
AK302435.1 | - | 4 - 5 | 450 | 97 | |
BC146822.1 | - | 4 - 5 | 466 | 97 |