Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024513.3 | NP_078789.2 | 14 - 15 | 4153 | 75 |
XM_006713333.3 | XP_006713396.1 | 15 - 16 | 5318 | 75 | |
XM_006713334.3 | XP_006713397.1 | 14 - 15 | 5234 | 75 | |
XM_011534111.2 | XP_011532413.1 | 15 - 16 | 4231 | 75 | |
GenBank mRNA | AJ292348.2 | - | 14 - 15 | 4137 | 75 |
AK074165.1 | - | 2 - 3 | 304 | 75 | |
AL832358.1 | - | 14 - 15 | 4141 | 75 | |
AL833308.2 | - | 7 - 8 | 2139 | 75 | |
BC101468.1 | - | 13 - 14 | 4023 | 75 | |
BC101470.1 | - | 13 - 14 | 4023 | 75 |