Assay Design



Gene Transcripts

Gene Symbol
FYCO1
Entrez Gene ID
79443
Gene Name
FYVE and coiled-coil domain containing 1
Gene Aliases
CATC2, CTRCT18, RUFY3, ZFYVE7
Chromosome Location
Chr.3: 45917899 - 45995824 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024513.3 NP_078789.2 4 - 5 510 102
XM_006713333.3 XP_006713396.1 5 - 6 1675 102
XM_006713334.3 XP_006713397.1 4 - 5 1591 102
XM_011534111.2 XP_011532413.1 5 - 6 588 102
GenBank mRNA AJ292348.2 - 4 - 5 494 102
AK023397.1 - 1 - 2 130 102
AL832358.1 - 4 - 5 498 102
BC007218.1 - 2 - 3 177 102
BC101468.1 - 3 - 4 380 102
BC101470.1 - 3 - 4 380 102
BC143368.1 - 3 - 4 380 102
BP249484.1 - 4 - 5 501 102

View Full Product Details