Assay Design



Gene Transcripts

Gene Symbol
FYCO1
Entrez Gene ID
79443
Gene Name
FYVE and coiled-coil domain containing 1
Gene Aliases
CATC2, CTRCT18, RUFY3, ZFYVE7
Chromosome Location
Chr.3: 45917899 - 45995824 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024513.3 NP_078789.2 8 - 9 3270 67
XM_006713333.3 XP_006713396.1 9 - 10 4435 67
XM_006713334.3 XP_006713397.1 8 - 9 4351 67
XM_011534111.2 XP_011532413.1 9 - 10 3348 67
GenBank mRNA AJ292348.2 - 8 - 9 3254 67
AL832358.1 - 8 - 9 3258 67
AL833308.2 - 1 - 2 1256 67
BC101468.1 - 7 - 8 3140 67
BC101470.1 - 7 - 8 3140 67
BC143368.1 - 7 - 8 3140 67

View Full Product Details