Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138715.2 | NP_619729.1 | 9 - 10 | 1412 | 144 |
GenBank mRNA | AK293217.1 | - | 9 - 10 | 1332 | 144 |
BC063878.1 | - | 9 - 10 | 1313 | 144 | |
D90187.1 | - | 9 - 10 | 1260 | 144 |