Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002445.3 | NP_002436.1 | 7 - 8 | 1180 | 61 |
NM_138715.2 | NP_619729.1 | 7 - 8 | 1180 | 61 | |
NM_138716.2 | NP_619730.1 | 7 - 8 | 1180 | 61 | |
GenBank mRNA | AF037351.1 | - | 6 - 7 | 982 | 61 |
AK293217.1 | - | 7 - 8 | 1100 | 61 | |
AK293409.1 | - | 5 - 6 | 513 | 61 | |
BC063878.1 | - | 7 - 8 | 1081 | 61 | |
D90187.1 | - | 7 - 8 | 1028 | 61 | |
D90188.1 | - | 7 - 8 | 1048 | 61 |