Assay Design



Gene Transcripts

Gene Symbol
C20orf85
Entrez Gene ID
128602
Gene Name
chromosome 20 open reading frame 85
Gene Aliases
LLC1, bA196N14.1
Chromosome Location
Chr.20: 58150927 - 58161130 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178456.2 NP_848551.1 2 - 3 191 105
GenBank mRNA BC035405.1 - 2 - 3 180 105
BX093883.1 - 2 - 3 191 105

View Full Product Details