Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000152.4 | NP_000143.2 | 2 - 3 | 942 | 66 |
NM_001079803.2 | NP_001073271.1 | 3 - 4 | 757 | 66 | |
NM_001079804.2 | NP_001073272.1 | 2 - 3 | 677 | 66 | |
XM_005257193.2 | XP_005257250.1 | 3 - 4 | 743 | 66 | |
XM_005257194.4 | XP_005257251.1 | 3 - 4 | 783 | 66 | |
GenBank mRNA | AK299462.1 | - | - | 721 | 66 |
BC040431.1 | - | 3 - 4 | 689 | 66 | |
BF206270.1 | - | 1 - 2 | 464 | 66 | |
DQ907243.1 | - | 1 - 2 | 545 | 66 | |
M34424.1 | - | 1 - 2 | 577 | 66 | |
Y00839.1 | - | 2 - 3 | 764 | 66 |