Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001316327.1 | NP_001303256.1 | 8 - 9 | 838 | 57 |
NM_006254.3 | NP_006245.2 | 8 - 9 | 1005 | 57 | |
NM_212539.1 | NP_997704.1 | 7 - 8 | 893 | 57 | |
XM_006713257.3 | XP_006713320.1 | 7 - 8 | 812 | 57 | |
XM_006713259.2 | XP_006713322.1 | 9 - 10 | 1109 | 57 | |
XM_017006855.1 | XP_016862344.1 | 7 - 8 | 690 | 57 | |
XM_017006856.1 | XP_016862345.1 | 7 - 8 | 1264 | 57 | |
GenBank mRNA | AK130150.1 | - | 3 - 4 | 196 | 57 |
AK294272.1 | - | 5 - 6 | 464 | 57 | |
AK313216.1 | - | 8 - 9 | 838 | 57 | |
BC043350.1 | - | 8 - 9 | 1024 | 57 | |
D10495.1 | - | 5 - 6 | 511 | 57 | |
L07860.1 | - | 7 - 8 | 710 | 57 | |
L07861.1 | - | 7 - 8 | 710 | 57 | |
Z22521.1 | - | 7 - 8 | 981 | 57 |