Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003076.4 | NP_003067.3 | 10 - 11 | 1441 | 70 |
XM_005269107.3 | XP_005269164.2 | 9 - 10 | 1318 | 70 | |
GenBank mRNA | AF109733.1 | - | 10 - 11 | 1229 | 70 |
AK130517.1 | - | 11 - 12 | 1181 | 70 | |
AK292700.1 | - | 10 - 11 | 1105 | 70 | |
BC009368.2 | - | 10 - 11 | 1391 | 70 |