Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005914.2 | NP_001005914.1 | 16 - 17 | 1988 | 91 |
NM_001290060.1 | NP_001276989.1 | 15 - 16 | 1945 | 91 | |
NM_001290061.1 | NP_001276990.1 | 15 - 16 | 1794 | 91 | |
NM_001290062.1 | NP_001276991.1 | 9 - 10 | 1211 | 91 | |
NM_001290063.1 | NP_001276992.1 | 9 - 10 | 1085 | 91 | |
NM_004636.3 | NP_004627.1 | 16 - 17 | 1991 | 91 | |
NR_110697.1 | 6 - 7 | 712 | 91 | ||
GenBank mRNA | AB083186.1 | - | 15 - 16 | 1741 | 91 |
AB209322.1 | - | 14 - 15 | 2171 | 91 | |
AF217991.1 | - | 15 - 16 | 1754 | 91 | |
AK092182.1 | - | 6 - 7 | 674 | 91 | |
AK098329.1 | - | 4 - 5 | 1820 | 91 | |
AK293678.1 | - | 9 - 10 | 1044 | 91 | |
BC009113.1 | - | 12 - 13 | 1695 | 91 | |
BC013975.2 | - | 15 - 16 | 1768 | 91 | |
BC024220.2 | - | 15 - 16 | 1768 | 91 | |
BC044602.1 | - | 9 - 10 | 1173 | 91 | |
U28369.1 | - | 15 - 16 | 1934 | 91 |