Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005914.2 | NP_001005914.1 | 5 - 6 | 742 | 81 |
NM_001290060.1 | NP_001276989.1 | 4 - 5 | 696 | 81 | |
NM_001290061.1 | NP_001276990.1 | 4 - 5 | 530 | 81 | |
NM_004636.3 | NP_004627.1 | 5 - 6 | 742 | 81 | |
GenBank mRNA | AB083186.1 | - | 4 - 5 | 477 | 81 |
AB209322.1 | - | 4 - 5 | 817 | 81 | |
AF217991.1 | - | 4 - 5 | 506 | 81 | |
AK308875.1 | - | 4 - 5 | 774 | 81 | |
BC009113.1 | - | 1 - 2 | 449 | 81 | |
BC013975.2 | - | 4 - 5 | 522 | 81 | |
BC024220.2 | - | 4 - 5 | 522 | 81 | |
U28369.1 | - | 4 - 5 | 685 | 81 |