Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190945.1 | NP_001177874.1 | 8 - 9 | 1455 | 94 |
NM_001190947.1 | NP_001177876.1 | 5 - 6 | 867 | 94 | |
NM_005658.4 | NP_005649.1 | 7 - 8 | 1602 | 94 | |
GenBank mRNA | AK090468.1 | - | 7 - 8 | 94 | |
AK295959.1 | - | 5 - 6 | 94 | ||
AK315476.1 | - | 8 - 9 | 94 | ||
AL832989.1 | - | 7 - 8 | 94 | ||
BC024145.2 | - | 6 - 7 | 94 | ||
BT019408.1 | - | 6 - 7 | 1030 | 94 | |
U19261.1 | - | 6 - 7 | 1105 | 94 |