Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004668.2 | NP_004659.2 | 20 - 21 | 2427 | 92 |
XM_006716168.2 | XP_006716231.1 | 20 - 21 | 2485 | 92 | |
XM_011516670.2 | XP_011514972.1 | 20 - 21 | 2493 | 92 | |
XM_011516671.2 | XP_011514973.1 | 21 - 22 | 2560 | 92 | |
XM_011516672.2 | XP_011514974.1 | 21 - 22 | 2546 | 92 | |
XM_011516673.2 | XP_011514975.1 | 20 - 21 | 2448 | 92 | |
XM_011516674.2 | XP_011514976.1 | 20 - 21 | 2485 | 92 | |
XM_017012772.1 | XP_016868261.1 | 21 - 22 | 2465 | 92 | |
GenBank mRNA | AF016833.2 | - | 20 - 21 | 2427 | 92 |
BC120872.1 | - | 20 - 21 | 2399 | 92 |