Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004668.2 | NP_004659.2 | 8 - 9 | 1042 | 99 |
XM_006716168.2 | XP_006716231.1 | 8 - 9 | 1100 | 99 | |
XM_011516670.2 | XP_011514972.1 | 8 - 9 | 1108 | 99 | |
XM_011516671.2 | XP_011514973.1 | 9 - 10 | 1175 | 99 | |
XM_011516672.2 | XP_011514974.1 | 9 - 10 | 1161 | 99 | |
XM_011516673.2 | XP_011514975.1 | 8 - 9 | 1063 | 99 | |
XM_011516674.2 | XP_011514976.1 | 8 - 9 | 1100 | 99 | |
XM_017012772.1 | XP_016868261.1 | 9 - 10 | 1080 | 99 | |
GenBank mRNA | AF016833.2 | - | 8 - 9 | 1042 | 99 |
BC120872.1 | - | 8 - 9 | 1014 | 99 |