Assay Design



Gene Transcripts

Gene Symbol
CAPS
Entrez Gene ID
828
Gene Name
calcyphosine
Gene Aliases
CAPS1
Chromosome Location
Chr.19: 5913644 - 5916211 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080590.3 NP_542157.2 4 - 5 1070 133
GenBank mRNA BC019065.1 - - 483 133

View Full Product Details