Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014352.3 | NP_055167.2 | 1 - 2 | 176 | 59 |
XM_011542741.2 | XP_011541043.1 | 1 - 2 | 135 | 59 | |
XM_017017487.1 | XP_016872976.1 | 1 - 2 | 176 | 59 | |
GenBank mRNA | AF162278.1 | - | 1 - 2 | 140 | 59 |
AK291993.1 | - | 1 - 2 | 176 | 59 | |
BC101649.1 | - | 1 - 2 | 111 | 59 | |
BC143593.1 | - | 1 - 2 | 111 | 59 | |
BC143594.1 | - | 1 - 2 | 111 | 59 | |
BC143595.1 | - | 1 - 2 | 109 | 59 |