Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244682.1 | NP_001231611.1 | 3 - 4 | 174 | 69 |
NM_014352.3 | NP_055167.2 | 3 - 4 | 284 | 69 | |
XM_011542739.2 | XP_011541041.1 | 5 - 6 | 499 | 69 | |
XM_011542740.2 | XP_011541042.1 | 4 - 5 | 357 | 69 | |
XM_017017487.1 | XP_016872976.1 | 3 - 4 | 284 | 69 | |
GenBank mRNA | AF162278.1 | - | 3 - 4 | 248 | 69 |
AJ012214.1 | - | 2 - 3 | 134 | 69 | |
AK291993.1 | - | 3 - 4 | 284 | 69 | |
AK302212.1 | - | 3 - 4 | 174 | 69 | |
BC101649.1 | - | 3 - 4 | 219 | 69 | |
BC143594.1 | - | 3 - 4 | 219 | 69 | |
BC143595.1 | - | 3 - 4 | 217 | 69 |