Assay Design



Gene Transcripts

Gene Symbol
POU2F3
Entrez Gene ID
25833
Gene Name
POU class 2 homeobox 3
Gene Aliases
Epoc-1, OCT-11, OCT11, OTF-11, PLA-1, PLA1, Skn-1a
Chromosome Location
Chr.11: 120236640 - 120319944 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244682.1 NP_001231611.1 6 - 7 487 108
NM_014352.3 NP_055167.2 6 - 7 597 108
XM_011542739.2 XP_011541041.1 8 - 9 812 108
XM_011542740.2 XP_011541042.1 7 - 8 670 108
XM_011542741.2 XP_011541043.1 5 - 6 521 108
XM_011542742.2 XP_011541044.1 7 - 8 775 108
XM_011542743.2 XP_011541045.1 5 - 6 2916 108
XM_017017487.1 XP_016872976.1 6 - 7 597 108
GenBank mRNA AF133895.1 - 6 - 7 454 108
AF162278.1 - 6 - 7 108
AJ012214.1 - 5 - 6 108
AK291993.1 - 6 - 7 108
AK302212.1 - 6 - 7 487 108
BC101649.1 - 6 - 7 532 108
BC143593.1 - 7 - 8 579 108
BC143594.1 - 7 - 8 576 108
BC143595.1 - 7 - 8 577 108

View Full Product Details