Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006383.3 | NP_006374.1 | 1 - 2 | 373 | 66 |
XM_005254126.3 | XP_005254183.1 | 1 - 2 | 814 | 66 | |
GenBank mRNA | AB012955.1 | - | 1 - 2 | 59 | 66 |
AK293167.1 | - | 1 - 2 | 287 | 66 | |
BC047381.1 | - | 1 - 2 | 301 | 66 | |
BC057292.1 | - | 1 - 2 | 301 | 66 | |
BE042882.1 | - | 1 - 2 | 109 | 66 | |
BQ681471.1 | - | - | 399 | 66 | |
BT019962.1 | - | 1 - 2 | 53 | 66 |