Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006383.3 | NP_006374.1 | 2 - 3 | 411 | 101 |
XM_005254126.3 | XP_005254183.1 | 2 - 3 | 852 | 101 | |
GenBank mRNA | AB012955.1 | - | 2 - 3 | 97 | 101 |
AK293167.1 | - | 2 - 3 | 325 | 101 | |
BC047381.1 | - | 2 - 3 | 339 | 101 | |
BC057292.1 | - | 2 - 3 | 339 | 101 | |
BE042882.1 | - | 2 - 3 | 147 | 101 | |
BQ681471.1 | - | - | 437 | 101 | |
BT019962.1 | - | 2 - 3 | 91 | 101 |