Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000408.4 | NP_000399.3 | 16 - 17 | 2194 | 168 |
NM_001083112.2 | NP_001076581.2 | 16 - 17 | 2420 | 168 | |
XM_005246469.2 | XP_005246526.1 | 16 - 17 | 2133 | 168 | |
XM_011510977.2 | XP_011509279.1 | 16 - 17 | 2188 | 168 | |
XM_011510978.2 | XP_011509280.1 | 17 - 18 | 2238 | 168 | |
XM_011510980.2 | XP_011509282.1 | 11 - 12 | 1419 | 168 | |
XM_017003830.1 | XP_016859319.1 | 16 - 17 | 2274 | 168 | |
XM_017003831.1 | XP_016859320.1 | 15 - 16 | 2083 | 168 | |
GenBank mRNA | AB209399.1 | - | 16 - 17 | 2189 | 168 |
AF311325.1 | - | 17 - 18 | 2230 | 168 | |
AK093198.1 | - | 17 - 18 | 2229 | 168 | |
AK290057.1 | - | 16 - 17 | 2355 | 168 | |
AK291065.1 | - | 16 - 17 | 2433 | 168 | |
AK292817.1 | - | 16 - 17 | 2193 | 168 | |
AK299645.1 | - | 12 - 13 | 1535 | 168 | |
BC019874.1 | - | 11 - 12 | 1348 | 168 | |
U12424.1 | - | 16 - 17 | 2183 | 168 | |
U36310.1 | - | 16 - 17 | 2163 | 168 |