Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083112.2 | NP_001076581.2 | 1 - 2 | 346 | 68 |
GenBank mRNA | AB209399.1 | - | 1 - 2 | 115 | 68 |
AK290057.1 | - | 1 - 2 | 281 | 68 | |
AK291065.1 | - | 1 - 2 | 359 | 68 | |
U36310.1 | - | 1 - 2 | 89 | 68 | |
U79250.1 | - | 1 - 2 | 368 | 68 |