Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032445.2 | NP_115821.2 | 20 - 21 | 2851 | 113 |
XM_017022672.1 | XP_016878161.1 | 19 - 20 | 3280 | 113 | |
GenBank mRNA | AB058677.1 | - | 18 - 19 | 2644 | 113 |
AB300051.1 | - | 19 - 20 | 2710 | 113 | |
BC029999.1 | - | 20 - 21 | 2883 | 113 | |
BC117419.1 | - | 20 - 21 | 2771 | 113 | |
BC126313.1 | - | 20 - 21 | 2771 | 113 |