Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320273.1 | NP_001307202.1 | 1 - 2 | 284 | 82 |
NM_001320274.1 | NP_001307203.1 | 1 - 2 | 284 | 82 | |
NM_030965.2 | NP_112227.1 | 1 - 2 | 284 | 82 | |
GenBank mRNA | AK056241.1 | - | 1 - 2 | 251 | 82 |
AK095758.1 | - | 1 - 2 | 212 | 82 | |
AK295019.1 | - | 1 - 2 | 200 | 82 | |
AK300902.1 | - | 1 - 2 | 209 | 82 | |
BC001201.1 | - | 1 - 2 | 191 | 82 | |
DA166913.1 | - | 1 - 2 | 243 | 82 | |
DC358960.1 | - | 1 - 2 | 284 | 82 |