Assay Design



Gene Transcripts

Gene Symbol
ST6GALNAC5
Entrez Gene ID
81849
Gene Name
ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 5
Gene Aliases
SIAT7-E, SIAT7E, ST6GalNAcV
Chromosome Location
Chr.1: 76867408 - 77069167 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030965.2 NP_112227.1 2 - 3 529 77
GenBank mRNA AJ507292.1 - 2 - 3 273 77
AK056241.1 - 2 - 3 496 77
AK095758.1 - 3 - 4 419 77
AK295019.1 - 3 - 4 372 77
BC001201.1 - 2 - 3 436 77

View Full Product Details