Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167595.1 | NP_001161067.1 | 1 - 2 | 343 | 66 |
NM_014324.5 | NP_055139.4 | 1 - 2 | 343 | 66 | |
NM_203382.2 | NP_976316.1 | 1 - 2 | 343 | 66 | |
GenBank mRNA | AF047020.1 | - | 1 - 2 | 335 | 66 |
AF158378.1 | - | 1 - 2 | 312 | 66 | |
AJ130733.1 | - | 1 - 2 | 333 | 66 | |
AK000912.1 | - | 1 - 2 | 328 | 66 | |
AK022765.1 | - | 1 - 2 | 313 | 66 | |
AK222831.1 | - | 1 - 2 | 315 | 66 | |
AK225749.1 | - | 1 - 2 | 352 | 66 | |
AK292988.1 | - | 1 - 2 | 293 | 66 | |
AL545355.3 | - | 1 - 2 | 256 | 66 | |
AY330488.1 | - | 1 - 2 | 256 | 66 | |
AY330489.1 | - | 1 - 2 | 256 | 66 | |
AY935981.1 | - | 1 - 2 | 256 | 66 | |
BC009471.1 | - | 1 - 2 | 259 | 66 | |
BG708726.1 | - | 1 - 2 | 279 | 66 | |
BI550368.1 | - | 1 - 2 | 381 | 66 | |
BT007193.1 | - | 1 - 2 | 247 | 66 | |
DA156454.1 | - | 1 - 2 | 346 | 66 | |
EF560721.1 | - | 1 - 2 | 266 | 66 | |
FJ498906.1 | - | 1 - 2 | 259 | 66 | |
FJ498907.1 | - | 1 - 2 | 259 | 66 | |
FJ498908.1 | - | 1 - 2 | 259 | 66 |