Assay Design
Gene Transcripts
- Gene Symbol
- SRSF2
- Entrez Gene ID
-
6427
- Gene Name
- serine and arginine rich splicing factor 2
- Gene Aliases
-
PR264, SC-35, SC35, SFRS2, SFRS2A, SRp30b
- Chromosome Location
-
Chr.17: 76734115 - 76737431 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC066958.1 |
- |
4 - 5 |
903 |
101 |
View Full Product Details