Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079803.2 | NP_001073271.1 | 1 - 2 | 100 | 125 |
NR_134848.1 | 1 - 2 | 100 | 125 | ||
XM_005257194.4 | XP_005257251.1 | 1 - 2 | 91 | 125 | |
GenBank mRNA | HY101855.1 | - | 1 - 2 | 100 | 125 |