Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321173.1 | NP_001308102.1 | 3 - 4 | 390 | 106 |
NM_005486.2 | NP_005477.2 | 3 - 4 | 390 | 106 | |
GenBank mRNA | AJ010071.1 | - | 3 - 4 | 245 | 106 |
AK223125.1 | - | 3 - 4 | 224 | 106 | |
AK225351.1 | - | 3 - 4 | 242 | 106 | |
AK303912.1 | - | 2 - 3 | 144 | 106 | |
AK315039.1 | - | 3 - 4 | 228 | 106 | |
BC029396.1 | - | 3 - 4 | 224 | 106 |