Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135651.2 | NP_001129123.1 | 2 - 3 | 301 | 91 |
NM_002759.3 | NP_002750.1 | 2 - 3 | 535 | 91 | |
XM_011532987.2 | XP_011531289.1 | 2 - 3 | 235 | 91 | |
XM_017004503.1 | XP_016859992.1 | 2 - 3 | 231 | 91 | |
GenBank mRNA | AK290655.1 | - | 2 - 3 | 221 | 91 |
AK313818.1 | - | 1 - 2 | 58 | 91 | |
BC007769.2 | - | 1 - 2 | 77 | 91 | |
BC022314.1 | - | 2 - 3 | 189 | 91 | |
BC040851.1 | - | 2 - 3 | 301 | 91 | |
BC057805.1 | - | 2 - 3 | 206 | 91 | |
M35663.1 | - | 1 - 2 | 164 | 91 |