Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135651.2 | NP_001129123.1 | 5 - 6 | 710 | 149 |
NM_001135652.2 | NP_001129124.1 | 3 - 4 | 403 | 149 | |
NM_002759.3 | NP_002750.1 | 5 - 6 | 944 | 149 | |
XM_011532987.2 | XP_011531289.1 | 5 - 6 | 644 | 149 | |
XM_017004503.1 | XP_016859992.1 | 5 - 6 | 640 | 149 | |
GenBank mRNA | AK290655.1 | - | 5 - 6 | 630 | 149 |
AK313818.1 | - | 4 - 5 | 467 | 149 | |
AY302136.1 | - | 3 - 4 | 387 | 149 | |
BC007769.2 | - | 4 - 5 | 486 | 149 | |
BC022314.1 | - | 5 - 6 | 598 | 149 | |
BC040851.1 | - | 5 - 6 | 710 | 149 | |
BC057805.1 | - | 5 - 6 | 615 | 149 | |
BC093676.1 | - | 3 - 4 | 415 | 149 | |
BC101475.1 | - | 3 - 4 | 415 | 149 | |
BC143228.1 | - | 3 - 4 | 414 | 149 | |
BC143230.1 | - | 3 - 4 | 415 | 149 | |
M35663.1 | - | 4 - 5 | 573 | 149 | |
M85294.1 | - | 3 - 4 | 417 | 149 |