Assay Design
Gene Transcripts
- Gene Symbol
- SRSF6
- Entrez Gene ID
-
6431
- Gene Name
- serine and arginine rich splicing factor 6
- Gene Aliases
-
B52, HEL-S-91, SFRS6, SRP55
- Chromosome Location
-
Chr.20: 43457864 - 43463605 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_034009.1 |
|
2 - 3 |
427 |
70 |
GenBank mRNA |
AK300411.1 |
- |
2 - 3 |
395 |
70 |
U30828.1 |
- |
2 - 3 |
362 |
70 |
View Full Product Details