Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142725.1 | NP_001136197.1 | 5 - 6 | 499 | 101 |
NM_080663.2 | NP_542394.2 | 5 - 6 | 499 | 101 | |
GenBank mRNA | AK127391.1 | - | 5 - 6 | 502 | 101 |
AK292594.1 | - | 6 - 7 | 712 | 101 | |
BC010503.1 | - | 5 - 6 | 499 | 101 | |
BX437401.2 | - | 2 - 3 | 188 | 101 |